missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TK1, Human, Matched Antibody Pair (AP41), Abnova™
Description
Includes
Capture antibody: mouse monoclonal anti-TK1, IgG2a Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-TK1, IgG2b Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
Specifications
| Antigen | TK1 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Biotin |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TK1. |
| Gene | TK1 |
| Gene Alias | TK2 |
| Gene Symbols | TK1 |
| Host Species | Mouse |
| Quantity | 1 Set |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?