missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526133
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, non-human primate, rat and rabbit based on sequence homology.
TIMP3 is one of inhibitors of the matrix metalloproteinases, a group of peptidases involved in degradation of the extracellular matrix (ECM).
Specifications
| TIMP3 | |
| Polyclonal | |
| Unconjugated | |
| P35625, P39876 | |
| TIMP3 | |
| KLH conjugated synthetic peptide between 113-139 amino acids from the central region of human TIMP3 | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| TIMP3 | |
| HSMRK222, K222, K222TA2, SFD, MIG-5 protein, TIMP-3, metalloproteinase inhibitor 3, protein MIG-5, tissue inhibitor of metalloproteinases 3 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 21859, 7078 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction