Learn More
acyl-CoA thioesterase 13, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ACOT13 protein.
Brand: Abnova H00055856-B01.50uL
Description
This gene encodes a member of the thioesterase superfamily. In humans, the protein co-localizes with microtubules and is essential for sustained cell proliferation. The orthologous mouse protein forms a homotetramer and is associated with mitochondria. The mouse protein functions as a medium- and long-chain acyl-CoA thioesterase. Multiple transcript variants encoding different isoforms have been found for this gene
Sequence: MTSMTQSLREVIKAMTKARNFERVLGKITLVSAAPGKVICEMKVEEEHTNAIGTLHGGLTATLVDNISTMALLCTERGAPGVSVDMNITYMSPAKLGEDIVITAHVLKQGKTLAFTSVDLTNKATGKLIAQGRHTKHLGNSpecifications
| acyl-CoA thioesterase 13 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ACOT13 protein. | |
| THEM2 | |
| HT012/MGC4961/PNAS-27 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 55856 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_018473.2 | |
| THEM2 | |
| ACOT13 (NP_060943.1, 1 a.a. ∼ 140 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.