missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TH1L Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519407
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, mouse and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
The NELF complex of proteins interacts with the DSIF protein complex to repress transcriptional elongation by RNA polymerase II. The protein encoded by this gene is an essential part of the NELF complex. Alternative translation initiation site usage results in the formation of two isoforms with different N-termini.
Specifications
| TH1L | |
| Polyclonal | |
| Unconjugated | |
| NELFCD | |
| 2410003I03Rik; C77797; HSPC130; Negative elongation factor C/D; negative elongation factor complex member C/D; negative elongation factor complex member C/D, Th1l; negative elongation factor complex member C/D,Th1l; negative elongation factor D; negative elongation factor D-like; negative elongation factor proteins C and D; NELF-C; NELF-C/D; NELFCD; Nelfd; NELF-D; TH1; TH1 drosophila homolog; TH1 homolog; TH1L; TH1-like; TH1-like homolog; TH1-like protein; trihydrophobin 1; trihydrophobin 1 homolog; zgc:55482 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 51497 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8IXH7 | |
| NELFCD | |
| Synthetic peptide sequence (NKRVSINKDE) corresponding to the internal amino acids of TH1L (aa 373-383). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction