missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TGN46 Polyclonal specifically detects TGN46 in Human, Mouse, Bovine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Flow (Intracellular), Knockout Validated.
Specifications
Specifications
| Antigen | TGN46 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, Immunoassay |
| Classification | Polyclonal |
| Concentration | 1.0 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2ug/mL, Flow Cytometry, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31455601), Immunohistochemistry-Paraffin 1:400, Immunoblotting reported in scientific literature (PMID 25410859), Flow (Intracellular), Knockout Validated |
| Formulation | PBS and 30% Glycerol with 0.05% Sodium Azide |
| Gene Accession No. | O43493 |
| Gene Alias | TGN 46, TGN 48, TGN 51, TGN38, TGN-38, TGN38 homolog, TGN46, TGN-46, TGN48, TGN51, TGOLN 2, TGOLN2, trans golgi network 38, trans golgi network 46, Trans Golgi network integral membrane protein 2, Trans Golgi network integral membrane protein 2 [Precursor], Trans Golgi network protein (46 48 51kD isoforms), Trans golgi network protein 2, Trans Golgi network protein TGN51, Trans-Golgi network integral membrane protein 2, Trans-Golgi network protein 2, Trans-Golgi network protein TGN51, TTGN 2, TTGN2 |
| Gene Symbols | TGOLN2 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?