Learn More
TGFB-induced factor homeobox 2, Mouse, Clone: 6A8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TGIF2.
Brand: Abnova H00060436-M06.100ug
Description
The protein encoded by this gene is a DNA-binding homeobox protein and a transcriptional repressor. The encoded protein appears to repress transcription by recruiting histone deacetylases to TGF beta-responsive genes. This gene is amplified and overexpressed in some ovarian cancers, and mutations in this gene can cause holoprosencephaly. [provided by RefSeq
Sequence: SMPLHSGQGEKPAAPFPRGELESPKPLVTPGSTLTLLTRAEAGSPTGGLFNTPPPTPPEQDKEDFSSFQLLVEVALQRAAEMELQKQQDPSLPLLHTPIPLVSENPSpecifications
| TGFB-induced factor homeobox 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_021809 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 60436 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Immunofluorescence, Western Blot | |
| 6A8 | |
| Mouse monoclonal antibody raised against a partial recombinant TGIF2. | |
| TGIF2 | |
| TGIF2 | |
| TGIF2 (NP_068581, 131 a.a. ∼ 236 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.