missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-TGF-beta RI/ALK-5, APC, Clone: 141231, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
Specifications
| Antigen | TGF-beta RI/ALK-5 |
|---|---|
| Clone | 141231 |
| Immunogen | S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TGF-beta RI/ALK-5 Ala21-Glu121 Accession # BAA05023 |
| Host Species | Rat |
| Primary or Secondary | Primary |
Description
TGF-beta RI/ALK-5 Monoclonal specifically detects TGF-beta RI/ALK-5 in Mouse samples. It is validated for Flow Cytometry.Specifications
| TGF-beta RI/ALK-5 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TGF-beta RI/ALK-5 Ala21-Glu121 Accession # BAA05023 | |
| Primary | |
| Mouse | |
| APC | |
| 7046 | |
| Purified | |
| Flow Cytometry | |
| RUO |
| 141231 | |
| Rat | |
| Monoclonal | |
| IgG2a | |
| AAT5, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, Activin receptor-like kinase 5, ACVRLK4, ALK-5, ALK-5ALK5, EC 2.7.11, EC 2.7.11.30, LDS1A, LDS2A, Serine/threonine-protein kinase receptor R4, SKR4, TbetaR-I, TGFB1R1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, TGFbetaRI, TGFBR1, TGF-bRI, TGFR-1, transforming growth factor beta receptor I, transforming growth factor, beta receptor 1, transforming growth factor, beta receptor I (activin A receptor type II-likekinase, 53kD), Transforming growth factor-beta receptor type I | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Protein A or G purified from hybridoma culture supernatant | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title