Learn More
TGF beta-1 Mouse anti-Mouse, Clone: 9016, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523702
Description
In sandwich ELISAs, less than 2% cross-reactivity with recombit human (rh) TGF- beta 3 and recombit amphibian TGF- beta 5 and no cross-reactivity with recombit porcine TGF-beta 2 or rhTGF-beta 2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
TGF beta-1 (TGFB1, Transforming Growth Factor Beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its pro-domain. Activation of TGF beta-1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF beta-1, an amino-terminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP. TGF beta-1 helps regulates proliferation, differentiation, adhesion, migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. TGF beta-1 is cleaved into a latency-associated peptide and a mature TGF beta-1 peptide, and is found in either a latent form composed of a TGFB1 homodimer, a LAP homodimer, and a latent TGFB1-binding protein, or in an active form composed of a TGF beta-1 homodimer. The mature peptide may also form heterodimers with other TGF beta family members. The gene for TGF beta-1 is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease and cystic fibrosis.
Specifications
| TGF beta-1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P04202 | |
| TGFB1 | |
| CHO-derived recombit TGF-beta 1 and latent TGF-beta 1 | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Neutralization, Western Blot | |
| 9016 | |
| Unconjugated | |
| Tgfb1 | |
| TGFB, TGFb-1, DPD1, Transforming Growth Factor, Transforming Growth Factor Beta-1, TGF-beta-1, CED | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 21803 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.