missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-β1 Rat anti-Human, Pig, Unlabeled, Clone: A75-2, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 555052
This item is not returnable.
View return policy
Description
The A75-2 antibody reacts with mouse, pig, human TGF-β1. The immunogen used to generate this hybridoma was recombinant mouse TGF-β1.
ELISA, Western Blotting
Specifications
| TGF-β1 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Mouse TGF-β1 | |
| 0.5 mg | |
| Immunology | |
| Human, Mouse, Pig | |
| IgG2a κ |
| ELISA | |
| A75-2 | |
| Unconjugated | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction