Learn More
TFE3 Mouse anti-Mouse, Unlabeled, Clone: G138-312, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554263
Description
TFE3 is a ubiquitously expressed 50kDa transcription factor containing both basic helix-loop-helix (bHLH) and leucine zipper (ZIP) motifs. It was initially identified as a protein which bound to the μE3 site in the immunoglobulin heavy chain enhancer. In addition, it binds to the MLTF/USF site in the adenovirus major late promoter. Upon binding as a dimer, it induces a minor groove-oriented bend in the DNA, similar to other HLH proteins such as TFEB, USF, myc, and max. Binding results in transcriptional activation and is mediated through the basic region located just N-terminal to the HLH domain. Clone G138-312 recognizes TFE3. The antibody was raised against a bacterially expressed TFE3-L protein consisting of 327 amino acids encoded by nucleotides 361-2469 of the cloned cDNA.
Host Species: Mouse
Clone: G138-312
Isotype: IgG1
Species Reactivity: Human
Immunogen: TFE3-L Protein
Formula Weight [Chemical]: 50kDa
Immunoprecipitation, Western Blotting
Specifications
| TFE3 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| TFE3-L Protein | |
| 0.1 mg | |
| Cell Biology | |
| Human, Mouse | |
| IgG1 |
| Western Blot | |
| G138-312 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.