missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFB2M, Mouse, Clone: 2E10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TFB2M.
Brand: Abnova H00064216-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: YLIQMIPRQNLFTKNLTPMNYNIFFHLLKHCFGRRSATVIDHLRSLTPLDARDILMQIGKQEDEKVVNMHPQDFKTLFETIERSKDCAYKWLYDETLEDRSpecifications
| TFB2M | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_022366 | |
| TFB2M | |
| TFB2M (NP_071761, 297 a.a. ∼ 396 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 2E10 | |
| Mouse monoclonal antibody raised against a partial recombinant TFB2M. | |
| TFB2M | |
| FLJ22661/FLJ23182/Hkp1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 64216 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction