Learn More
transcription factor AP-4 (activating enhancer binding protein 4), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant TFAP4.
Brand: Abnova H00007023-A01.50uL
Description
Transcription factors of the basic helix-loop-helix-zipper (bHLH-ZIP) family contain a basic domain, which is used for DNA binding, and HLH and ZIP domains, which are used for oligomerization. Transcription factor AP4 activates both viral and cellular genes by binding to the symmetrical DNA sequence CAGCTG (Mermod et al., 1988 [PubMed 2833704]; Hu et al., 1990 [PubMed 2123466]).[supplied by OMIM
Sequence: AEYIFSLEQEKTRLLQQNTQLKRFIQELSGSSPKRRRAEDKDEGIGSPDIWEDEKAEDLRREMIELRQQLDKERSVRMMLEEQVRSLEAHMYPEKLKVIASpecifications
| transcription factor AP-4 (activating enhancer binding protein 4) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant TFAP4. | |
| TFAP4 | |
| AP-4/bHLHc41 | |
| Mouse | |
| 50 μL | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_003223 | |
| TFAP4 | |
| TFAP4 (NP_003214, 93 a.a. ∼ 192 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 7023 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.