missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBIAD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA146133
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The target sequence has 85% sequence homology with mouse and rat proteins. Immunogen sequence: PEQDRLPQRS WRQK.
A novel gene designated TERE1, has a significant effect on the growth regulation in bladder cancer. The TERE1 gene maps to a chromosome locus that has been identified by loss of heterozygosity studies as a site of a putative tumor suppressor gene or genes for multiple tumor types including prostate carcinoma. Data suggest that TERE1 may be significant in prostate cancer growth regulation and the down regulation or absence of TERE1 may be an important component of the phenotype of advanced disease. Data also suggest a potential role for this gene product in the progression of bladder cancer.
Spécification
| UBIAD1 | |
| Polyclonal | |
| Unconjugated | |
| UBIAD1 | |
| 1200002M06Rik; AI426463; AW320947; RGD1309588; RP4-796F18.1; SCCD; Tere1; transitional epithelia response protein; transitional epithelial response protein 1; UbiA prenyltransferase domain containing 1; ubiA prenyltransferase domain-containing protein 1; UBIAD1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29914 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9Y5Z9 | |
| UBIAD1 | |
| A synthetic peptide made to residues 32-45 of the 37 kDa human TERE1 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu