missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TER-119/Erythroid Cells Rat anti-Mouse, Unlabeled, Clone: TER-119, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 550565
This item is not returnable.
View return policy
Description
Specifications
| TER-119/Erythroid Cells | |
| Monoclonal | |
| 31.25μg/mL | |
| Lymphocyte antigen 76; Ly76; Ly-76; TER-119; Ter119 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| TER-119 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide. | |
| Mouse Fetal Liver | |
| 1 mL | |
| Immunology | |
| Mouse | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction