Learn More
TER-119/Erythroid Cells Rat anti-Mouse, Unlabeled, Clone: TER-119, BD
Rat Monoclonal Antibody
£178.33 - £325.79
Specifications
| Antigen | TER-119/Erythroid Cells |
|---|---|
| Clone | TER-119 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15867328
|
BD Biosciences
553670 |
0.1 mg |
£186.00 £178.33 / Each Save £7.67 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15877328
|
BD Biosciences
553671 |
0.5 mg |
£340.00 £325.79 / Each Save £14.21 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TER-119 antibody reacts with a 52kDa molecule associated with glycophorin A on cells of the erythroid lineage in embryonic yolk sac, fetal liver, newborn liver, adult bone marrow, adult peripheral blood, and adult lymphoid organs. The TER-119 antigen is expressed on erythroid cells from pro-erythroblast through mature erythrocyte stages, but not on cells with BFU-E or CFU-E activities. The TER-119 epitope is not detected on hematopoietic stem cells, lymphoid cells, myeloid cells, or erythroleukemia lines. The TER-119 mAb is a component of the “lineage” cocktail used in studies of hematopoietic progenitors to detect, or deplete cells committed to the hematopoietic lineages. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: TER-119
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: Mouse Fetal Liver
Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| TER-119/Erythroid Cells | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| Immunology | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2b κ | |
| Affinity Purified | |
| Lymphocyte antigen 76; Ly76; Ly-76; TER-119; Ter119 |
| TER-119 | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Mouse | |
| Mouse Fetal Liver | |
| Primary | |
| Store undiluted at 4°C |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.