Learn More
TER-119/Erythroid Cells Rat anti-Mouse, Biotin, Clone: TER-119, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553672
Description
The TER-119 antibody reacts with a 52kDa molecule associated with glycophorin A on cells of the erythroid lineage in embryonic yolk sac, fetal liver, newborn liver, adult bone marrow, adult peripheral blood, and adult lymphoid organs. The TER-119 antigen is expressed on erythroid cells from pro-erythroblast through mature erythrocyte stages, but not on cells with BFU-E or CFU-E activities. The TER-119 epitope is not detected on hematopoietic stem cells, lymphoid cells, myeloid cells, or erythroleukemia lines. The TER-119 mAb is a component of the “lineage cocktail” used in studies of hematopoietic progenitors to detect, or deplete cells committed to the hematopoietic lineages. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: TER-119
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: Mouse Fetal Liver
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| TER-119/Erythroid Cells | |
| Monoclonal | |
| 0.5mg/mL | |
| Lymphocyte antigen 76; Ly76; Ly-76; TER-119; Ter119 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| TER-119 | |
| Biotin | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse Fetal Liver | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.