Learn More
tektin 2 (testicular), Mouse, Clone: 2H4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TEKT2.
Brand: Abnova H00027285-M02.100ug
Description
This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. This gene is expressed in the testis and its protein is localized to the flagella of the sperms, indicating that it may play a role in spermatogenesis. [provided by RefSeq
Sequence: MATLSVKPSRRFQLPDWHTNSYLLSTNAQLQRDASHQIRQEARVLRNETNNQTIWDEHDNRTRLVERIDTVNRWKEMLDKCLTDLDAEIDALTQMKESASpecifications
| tektin 2 (testicular) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014466 | |
| TEKT2 | |
| TEKT2 (NP_055281, 1 a.a. ∼ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Adhesion | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2H4 | |
| Mouse monoclonal antibody raised against a partial recombinant TEKT2. | |
| TEKT2 | |
| TEKTB1/TEKTIN-T/h-tektin-t | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 27285 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.