Learn More
TCR V delta 2, FITC, clone: 15D, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen TCR2732
Description
The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
TCR2732 targets TCR V delta 2 in FACS, IHC(F), IP, and TCA applications and shows reactivity with Human samples.The TCR2732 immunogen is human TCR V delta 2.
Specifications
| TCR V delta 2 | |
| Monoclonal | |
| 0.1mg/mL | |
| TRD | |
| Mouse | |
| Protein A | |
| RUO | |
| 6964 | |
| Store at 4°C | |
| Purified |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, T-Cell Activation | |
| 15D | |
| FITC | |
| TRD | |
| Human TCR V delta 2 | |
| 200 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.