missing translation for 'onlineSavingsMsg'
Learn More

TCR gd Mouse anti-Human, Unlabeled, Clone: 11F2, BD

Product Code. 15886118
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15886118 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15886118 Supplier BD Biosciences Supplier No. 347900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The Anti–TCRγ/δ-1 antibody, clone 11F2, is derived from hybridization of mouse Sp2/0 myeloma cells with spleen cells from BALB/c mice immunized with a Sepharose™ bead/CD3/γ/δ TCR complex. Anti–TCRγ/δ-1 is mitogenic for γ/δ-TCR–bearing T lymphocytes.

The Anti–TCRγ/δ-1 antibody reacts with a framework epitope of the γ/δ T-cell antigen receptor (TCR). The γ/δ TCR is a heterodimeric glycoprotein that is noncovalently associated with the CD3 antigen. The γand δ TCR chains are composed of constant and variable regions, each encoded by distinct gene segments. The γchain forms either disulfide-linked or non–disulfide-linked heterodimers with the δ-subunit.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR gd
Applications Flow Cytometry
Classification Monoclonal
Clone 11F2
Concentration 25μg/mL
Conjugate Unconjugated
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Sepharose™ bead/CD3/γ/δ TCR complex
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.