Learn More
TCF-7/TCF-1 Mouse, PE, Clone: S33-966, BD
Description
The S33-966 monoclonal antibody specifically binds to TCF-7 (Transcription factor 7) which is also known as TCF-1 (T-cell factor 1). TCF-7 is a member of the High Mobility Group (HMG) DNA binding protein family of transcription factors. TCF-7 is expressed in thymocytes, T lymphocytes and proliferating epithelial cells. TCF-7 is expressed in the earliest T cell precursors and serves as a critical transcriptional regulator that is involved in the differentiation of thymocytes to become mature T lymphocytes. TCF-7 is necessary for the survival of immature CD4+CD8+ thymocytes. TCF-7 can also promote the differentiation of mature peripheral T cells to become Th2-like cells. TCF-7 may also function as a transcriptional repressor of certain target genes including CTNNB1 and TCF7L2 . The S33-966 antibody crossreacts with human TCF-7.
Specifications
Specifications
| Antigen | TCF-7/TCF-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | S33-966 |
| Concentration | 0.2mg/mL |
| Conjugate | PE |
| Description | TCF7; Transcription factor 7; TCF-1; TCF1; T-cell factor 1 |
| Host Species | Mouse |
| Immunogen | Mouse TCF-7 Recombinant Protein |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.