missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T-box 18, Mouse, Clone: 3C12, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant TBX18.
Brand: Abnova H00009096-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: STLLQGTGNGVPATHPHLLSGSSCSSPAFHLGPNTSQLCSLAPADYSACARSGLTLNRYSTSLAETYNRLTNQAGETFAPPRTPSYVGVSSSTSVNMSMGGTDGDTFSpecifications
| T-box 18 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| XM_496819 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 9096 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Western Blot | |
| 3C12 | |
| Mouse monoclonal antibody raised against a full length recombinant TBX18. | |
| TBX18 | |
| TBX18 | |
| TBX18 (XP_496819, 454 a.a. ∼ 560 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction