Learn More
anti-TBR1, Polyclonal, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Brand: Abcam AB31940-100UG
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Rat, Human
Immunogen: Synthetic peptide conjugated to KLH derived from within residues 50 - 150 of Mouse TBR1.
Applications:
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Specifications
| TBR1 | |
| 0.6-1mg/mL | |
| Rabbit | |
| Affinity Purified | |
| Neuroscience | |
| Human, Mouse, Rat |
| Polyclonal | |
| Unconjugated | |
| Synthetic peptide conjugated to KLH derived from within residues 50 - 150 of Mouse TBR1. | |
| 100 μg | |
| Primary | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.