missing translation for 'onlineSavingsMsg'
Learn More

anti-TBL1X, CTNNB1 Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0609.1Set

Product Code. 16131504

  • £585.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!


Includes: TBL1X rabbit purified polyclonal antibody (20μg); CTNNB1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols.
This item is not returnable. View return policy

Description

Description

Host Species: Rabbit, Mouse
Cross Reactivity: Human

In situ PLA

Specifications

Specifications

TBL1X
Monoclonal/Polyclonal
TBL1X
TBL1X
1 Set
6907/1499
Human
TBL1X & CTNNB1 Protein Protein Interaction Antibody Pair
In situ PLA
Unconjugated
EBI/SMAP55/TBL1
Mouse, Rabbit
Primary
Antibody pair set content: 1. TBL1X rabbit purified polyclonal antibody (20 ug) 2. CTNNB1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.