missing translation for 'onlineSavingsMsg'
Learn More

TBK1 (pS172) Mouse, Unlabeled, Clone: J133-1171, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 558397

Product Code. 15859348

  • ONLINE EXCLUSIVE PRICE
    £418.00
    £401.12 / Each
    Save £16.88
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NFκB is a ubiquitously expressed transcription factor that regulates many cytokine and Ig genes. It is involved in immune, inflammatory, viral, and acute phase responses. In most cells, NF-κB is sequestered in an inactive cytoplasmic form via interactions with the inhibitory proteins IkBα, IkBβ, and IkBε. Stimulation induces the release, activation, and nuclear translocation of NF-κB. Release of NF-κB results from the phosphorylation and proteolytic degradation of the IκB proteins. Two cytokine-inducible IκB kinases (IKKα and IKKβ) phosphorylate and target the IκB proteins for degradation via the ubiquitin pathway. IKKγ/NEMO, a third member of the IKK complex, functions as a regulatory subunit and interacts directly with IKKb. TBK1 (TANK-binding kinase 1, also known as T2K or NAK), a protein of 729 amino acids, is another member of the IKK family of kinases regulating NF-κB downstream of the tumor necrosis factor and Toll-like receptor pathways. TBK1 forms a complex with the adaptor proteins TANK (TRAF-associated NF-κB activator) and TRAF2 (TNF-receptor-associated factor 2), and this oligomer is required for activation and phosphorylation of TBK1 at serine 172 (S172). The J133-1171 monoclonal antibody recognizes the phosphorylated S172 of human TBK1.

Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TBK1 (pS172)
Monoclonal
0.5mg/mL
T2K, NAK
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Western Blot
J133-1171
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
Phosphorylated Human TBK1
0.1 mg
Cell Biology
Human, Mouse
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.