missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TBCK Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA534789

Product Code. 15338691

  • £478.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: HepG2, Molt-4, NIH-3T3, JC, BCL-1. Predicted reactivity: Mouse (100%), Rat (99%), Zebrafish (98%), Xenopus laevis (98%), Chicken (100%). IHC notes, Requires antigen retrieval using heat mediated 10mM Citrate buffer (pH6.0) or Tris-EDTA buffer (pH8.0) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Involved in the modulation of mTOR signaling and expression of mTOR complex components. Involved in the regulation of cell proliferation and growth. Involved in the control of actin-cytoskeleton organization.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TBCK
Polyclonal
Unconjugated
TBCK
1700120J03Rik; 9430001M19; A630047E20Rik; C030007I09Rik; HSPC302; IHPRF3; RGD1307816; TBC domain-containing protein kinase-like protein; TBC1 domain containing kinase; TBCK; TBCKL
Rabbit
Antigen affinity chromatography
RUO
271981, 93627
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.41 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q8BM85, Q8TEA7
TBCK
Recombinant protein encompassing a sequence within the center region of human TBCK. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.