Learn More
Tau Mouse anti-Rat, Unlabeled, Clone: TAU-5, BD
Description
Tau proteins are microtuble-associated proteins (MAPs) that are thought to function by stabilizing microtubles. They comprise a family of multiple polypeptides that originate from one gene. Alternate splicing of Tau mRNA and differential phosphorylation contribute to the heterogeneity of Tau. Typically, Tau proteins migrate between ∽45-70kDa with isoelectric points ranging from 6.5-8.0. Tau expression is widespread in the brain and is found in most cell types within the mammalian central nervous system. Tau has a complex biochemical nature that is partly due to its many isoforms. The number and molecular weights of Tau forms changes during brain development. Additionally, the molecular wieghts and forms of Tau proteins at a given point of development vary between species. Antibodies directed against different epitopes have been used to distinguish between Tau forms. The monoclonal antibody TAU-5 recognizes a phosphorylation-independent epitope on Tau proteins. The reduced molecular weight of Tau proteins may range between 45kDa to 95kDa. The TAU-5 antibody reacts with a phosphorylation-independent epitope in the middle of tau. It recognizes both the phosphorylated and non-phosphorylated form of Tau. The antibody has been characterized in rat tissue.
Host Species: Mouse
Clone: TAU-5
Isotype: IgG1
Species Reactivity: Rat
Immunohistochemistry, Western Blotting
Specifications
Specifications
| Antigen | Tau |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | TAU-5 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.