Learn More
Tau Monoclonal Antibody (HT7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MN1000
383.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : PN
Additional Details : Weight : 0.68040kg
Description
MN1000 recognizes normal Tau from human and bovine brain and PHF-Tau. No cross reactivity with Tau from rat brain has been observed. The epitope of this antibody has been mapped on human Tau between residue 159 and 163 (numbering according to human Tau40), corresponding to the amino acid sequence PPGQK. MN1000 detects Tau which has a predicted molecular weight of approximately 79 kDa. Purity is >95% as determined by SDS-PAGE.
Tau Monoclonal antibody specifically detects Tau in Bovine, Human samples. It is validated for ELISA, Immunohistochemistry (Free Floating), Immunohistochemistry (Paraffin), Western Blot.Specifications
Tau | |
Monoclonal | |
0.2 mg/mL | |
PBS with no preservative | |
P10636, P29172 | |
MAPT | |
Purified human Tau, epitope human Tau between residue 159 and 163 (numbering according to human Tau40), corresponding to the amino acid sequence PPGQK. | |
100 μg | |
Primary | |
Bovine, Human | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (Free Floating), Immunohistochemistry (Paraffin), Western Blot | |
HT7 | |
Unconjugated | |
MAPT | |
AI413597; AW045860; DDPAC; FLJ31424; FTDP17; FTDP-17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; Mapt; MAPTL; MGC138549; microtubule associated protein tau; microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; microtubules; MSTD; Mtapt; MTBT1; MTBT2; Neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; paired helical filament-tau; PHFtau; PHF-tau; PPND; PPP1R103; protein phosphatase 1, regulatory subunit 103; pTau; RNPTAU; Tau; Tau microtubule-associated protein; tau protein; Tau-4; Tau5; Unknown (protein for MGC:134287) | |
Mouse | |
Protein A | |
RUO | |
281296, 4137 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |