missing translation for 'onlineSavingsMsg'
Learn More

TAP Mouse, Unlabeled, Clone: 19, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611580

Product Code. 15805989

  • ONLINE EXCLUSIVE PRICE
    £198.00
    £190.20 / 1 set
    Save £7.80
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Nuclear export of cellular mRNAs is highly selective, wherein only fully processed RNAs are exported. Retroviral replication requires export of unspliced viral RNAs, which serve as templates for structural proteins and as genomic RNA for progeny virus. The export of unspliced viral RNA depends on the interaction of cis-acting RNA elements with cellular factors. In simian type D retroviruses, the cis-acting element is the constitutive transport element (CTE). CTE-dependent nuclear transport is mediated by the cellular TAP protein. TAP (Tip associating protein) is the vertebrate homolog of the yeast nuclear export protein Mex67p. TAP is a nuclear protein that shuttles between the nucleus and cytoplasm. It contains an N-terminal nuclear localization and export region (NLS-NES), a central RNA binding domain (RBD), and a C-terminal portion that is required for nuclear localization and nuclear rim association. The first 372 aa of TAP are necessary and sufficient for binding and nuclear export of CTE-containing RNA. Thus, TAP contains a novel RNA-binding motif that recognizes CTE-containing RNA and interacts with other components of the nuclear transport machinery.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TAP
Monoclonal
250μg/mL
Manganese Superoxide Dismutase
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
19
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human TAP aa. 488-596
50 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.