Learn More
TRAF family member-associated NFKB activator, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human TANK protein.
Brand: Abnova H00010010-D01P.100ug
Description
The TRAF (tumor necrosis factor receptor-associated factor) family of proteins associate with and transduce signals from members of the tumor necrosis factor receptor superfamily. The protein encoded by this gene is found in the cytoplasm and can bind to TRAF1, TRAF2, or TRAF3, thereby inhibiting TRAF function by sequestering the TRAFs in a latent state in the cytoplasm. For example, the protein encoded by this gene can block TRAF2 binding to LMP1, the Epstein-Barr virus transforming protein, and inhibit LMP1-mediated NF-kappa-B activation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MDKNIGEQLNKAYEAFRQACMDRDSAVKELQQKTENYEQRIREQQEQLSLQQTIIDKLKSQLLLVNSTQDNNYGCVPLLEDSETRKNNLTLDQPQDKVISGIAREKLPKVDIASAESSISpecifications
| TRAF family member-associated NFKB activator | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human TANK protein. | |
| TANK | |
| I-TRAF/TRAF2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10010 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_133484.1 | |
| TANK | |
| TANK (NP_597841.1, 1 a.a. ∼ 119 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.