Learn More
anti-TANK Matched Antibody Pair, Abnova
Brand: Abnova H00010010-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-TANK (100μg); Mouse polyclonal anti-TANK (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
The TRAF (tumor necrosis factor receptor-associated factor) family of proteins associate with and transduce signals from members of the tumor necrosis factor receptor superfamily. The protein encoded by this gene is found in the cytoplasm and can bind to TRAF1, TRAF2, or TRAF3, thereby inhibiting TRAF function by sequestering the TRAFs in a latent state in the cytoplasm. For example, the protein encoded by this gene can block TRAF2 binding to LMP1, the Epstein-Barr virus transforming protein, and inhibit LMP1-mediated NF-κ-B activation. Two transcript variants encoding different isoforms have been found for this gene.
ELISA
Specifications
| TANK | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TANK. | |
| I-TRAF/TRAF2 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-TANK (100 μg) 2. Detection antibody: mouse polyclonal anti-TANK (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| TANK | |
| TANK | |
| Affinity Purified | |
| RUO | |
| 10010 | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.