Learn More
TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human TAF7 protein.
Brand: Abnova H00006879-D01P.100ug
Description
The intronless gene for this transcription coactivator is located between the protocadherin beta and gamma gene clusters on chromosome 5. The protein encoded by this gene is a component of the TFIID protein complex, a complex which binds to the TATA box in class II promoters and recruits RNA polymerase II and other factors. This particular subunit interacts with the largest TFIID subunit, as well as multiple transcription activators. The protein is required for transcription by promoters targeted by RNA polymerase II. [provided by RefSeq
Sequence: MSKSKDDAPHELESQFILRLPPEYASTVRRAVQSGHVNLKDRLTIELHPDGRHGIVRVDRVPLASKLVDLPCVMESLKTIDKKTFYKTADICQMLVSTVDGDLYPPVEEPVASTDPKASKKKDKDKEKKFIWNHGITLPLKNVRKRRFRKTAKKKYIESPDVEKEVKRLLSTDAEAVSTRWEIIAEDETKEAENQGLDISSPGMSGHRQGHDSLEHDELREIFNDLSSSSEDEDETQHQDEEDINIIDTEEDLERQLQDKLNESDEQHQENEGTNQLVMGIQKQIDNMKGKLQETQDRAKRQEDLIMKVENLALKNRFQAVLDELKQKEDREKEQLSSLQEELESLLEKSpecifications
| TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human TAF7 protein. | |
| TAF7 | |
| TAF2F/TAFII55 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6879 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005642.2 | |
| TAF7 | |
| TAF7 (NP_005633.2, 1 a.a. ∼ 349 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.