missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-T- and B- Cell Activation Antigen; Clone: GL7; BD
Brand: BD Biosciences 562967
This item is not returnable.
View return policy
Description
Host Species: Rat
Clone GL7
Isotype IgM, κ
Species Reactivity Ms
Immunogen In vitro-activated T-cell-depleted CBA/Ca mouse splenocytes
Flow Cytometry
Specifications
| T- and B- Cell Activation Antigen | |
| GL7 | |
| Brilliant Violet 421 | |
| In vitro-activated T-cell-depleted CBA/Ca mouse splenocytes | |
| 50 μg | |
| Primary | |
| IgM |
| Monoclonal | |
| 0.2mg/mL | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction