Learn More
T- and B-Cell Activation Antigen Rat anti-Mouse, Alexa Fluor 647, Clone: GL7, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 561529
Description
The GL7 antibody reacts with a 35-kDa cell-surface protein found on T and B lymphocytes activated in vitro,on bone marrow Pre-B-II cells, germinal-center B cells, and a subpopulation of the thymocyte fraction expressing high density of CD3e. There is strain variability with respect to antigen distribution on thymocytes and Con A-activated spleen cells, with expression in BALB/c greater than that in C57BL/6.
Specifications
| T- and B-Cell Activation Antigen | |
| Monoclonal | |
| 0.2mg/mL | |
| GL7 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgM κ |
| Flow Cytometry | |
| GL7 | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| In vitro-activated T-cell-depleted CBA/Ca mouse splenocytes | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.