Learn More
α-Synuclein (pY125) Mouse anti-Human, Unlabeled, Clone: I57-628, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558246
Description
The 140-amino-acid α -Synuclein protein is identical to the non-amyloid- β component precursor (NACP), a presynaptic protein involved in amyloidogenesis in Alzheimer's disease (AD). This protein is expressed in brain, primarily in presynaptic nerve terminals. Although the exact function of the Synucleins has not been determined, they have been linked to the prominent neurodegenerative disorders AD and Parkinson's disease. The Tyrosine 125 (Y125) residue of α -Synuclein plays an important role in stress-induced dimerization of the protein and is phosphorylated by Pyk/RAFTK via the Src-family kinases Fyn and c-Src.
The I57-628 antibody recognizes α -Synuclein phosphorylated at Y125.
Specifications
| α-Synuclein (pY125) | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Phosphorylated peptide corresponding to the region including the Tyrosine 125 residue of α-Synuclein | |
| 0.1 mg | |
| Cell Biology | |
| Human | |
| IgG2b κ |
| Western Blot | |
| I57-628 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.