missing translation for 'onlineSavingsMsg'
Learn More

Syndecan-4 Rat anti-Mouse, Unlabeled, Clone: KY/8.2, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 550350

Product Code. 15816248

  • ONLINE EXCLUSIVE PRICE
    £119.00
    £114.13 / Each
    Save £4.87
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The KY/8.2 mAb reacts with mouse syndecan-4, a cell-surface heparan sulfate proteoglycan. Although the ligand(s) for this molecule is unknown, syndecans are known to interact with the heparin-binding domain of growth factors, chemokines, and extracellular matrix components. Accordingly, heparin inhibits the binding of syndecan-4 to the pre-B cell line BC7.12. Syndecan-4 is localized to focal adhesions and binds protein-kinase Cα. Thus, it is involved in cytoskeletal organization and signaling. Syndecan-4 is expressed on stromal cells and B-lymphocyte precursors in bone marrow, in addition to most splenic B cells and a small population of T lymphocytes. It is highly expressed on activated B lymphocytes, but is downregulated in isotype-switched memory B cells. The ectodomain of syndecan-4 is shed from cell surfaces constitutively and in response to specific physiological signals, and the resulting soluble fragments are functional. Immobilized KY/8.2 mAb induced the formation of dendritic processes on syndecan-4-transfected cells and on anti-CD40-activated splenic B cells. Other methods of activating B cells have not produced the same effect. This antibody recognizes an epitope in the N-terminus of syndecan-4, regardless of post-translational modifications.

Host Species: Rat
Clone: KY/8.2
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: BCB10 mouse pre-B cell line

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Syndecan 4
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
BCB10 mouse pre-B cell line
0.1 mg
Immunology
Mouse
IgG2a κ
Flow Cytometry
KY/8.2
Unconjugated
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.