Learn More
Syk Mouse anti-Human, FITC, Clone: 4D10, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 552476
Description
Syk is a member of the family of non-receptor type protein-tyrosine kinases and plays a crucial role in lymphocyte signaling and development. Syk is expressed in all haematopoietic cells and contributes to the signal transduction process by binding to a tyrosine-based activation motif (ITAM) of immune receptors, including Igα, TCRζ, CD3ε, FcεRIβ, and FceRIγ. Upon receptor activation, Syk binds to phosphorylated ITAMs via its two N-terminal SH2 domains thereby activating Syk and causing tyrosines in Syk to undergo auto-phosphorylation or phosphorylation. These phosphorylated sites can act as binding sites for other signaling molecules or help to regulate enzymatic activity. For example, in mast cells, Syk can activate downstream targets such as phospholipase Cγ1 and VAV. Studies in Syk-/- mast cells identified defects in both the ERK-MAP and JNK-MAP kinase pathways. Syk migrates as 72kDa in SDS/PAGE. (Predicted MW from SWISS-PROT is 72kDa; Accession number P43405).
Host Species: Mouse
Clone: 4D10
Isotype: IgG2a κ
Species Reactivity: Human
Immunogen: Human Syk peptide aa. 314-339
Flow Cytometry
Specifications
| Syk | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human Syk peptide aa. 314-339 | |
| 100 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 4D10 | |
| FITC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.