missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syk Mouse anti-Human, Clone: 8H5, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517206
This item is not returnable.
View return policy
Description
A suggested positive control for this product is THP-1 cells.
Syk, Spleen tyrosine kinase, is expressed in a wide range of hematopoietic and non-hematopoietic cells and plays a key role in adaptive immune system, which is the immunoreceptor (B cell receptor and Fc-receptors) signaling pathways including Fcgamma receptor-mediated phagocytosis. In hematopoietic cells such as B and T lymphocytes, natural killer cells, mast cells, macrophages and platelets, Syk is involved in the proximal signaling downstream of activated immunoreceptors (e.g. BCR, TCR, Fc receptors) containing immunoreceptor tyrosine-based activation motifs (ITAMs) to which it binds using its tandem SH2 domains. Activated and autophosphorylated Syk then phosphorylates its specific substrates including other enzymes and adaptor proteins, orchestrating a complex series of cellular responses such as cell proliferation, differentiation, survival and phagocytosis.
Specifications
| SYK | |
| Monoclonal | |
| Unconjugated | |
| SYK | |
| Tyrosine-protein kinase SYK; Spleen tyrosine kinase; p72-syk | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| 6850 | |
| -20°C | |
| Liquid |
| ELISA, Immunoprecipitation | |
| 8H5 | |
| HEPES with 0.01% BSA, 0.15M NaCl, 50% glycerol and 0.03% sodium azide | |
| P43405 | |
| SYK | |
| Synthetic peptide | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction