missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SURF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534228
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Mouse, Rat, Bovine, Turkey, Chicken, Xenopus, Salmon (100%).
This gene is located in the surfeit gene cluster, which is comprised of very tightly linked housekeeping genes that do not share sequence similarity. The encoded protein is a conserved integral membrane protein containing multiple putative transmembrane regions. In eukaryotic cells, protein transport between the endoplasmic reticulum and Golgi compartments is mediated in part by non-clathrin-coated vesicular coat proteins (COPs). The specific function of this protein has not been determined but its yeast homolog is directly required for packaging glycosylated pro-alpha-factor into COPII vesicles. This gene uses multiple polyadenylation sites, resulting in transcript length variation. The existence of alternatively spliced transcript variants has been suggested, but their validity has not been determined.
Specifications
| SURF4 | |
| Polyclonal | |
| Unconjugated | |
| SURF4 | |
| AL033340; AL033373; ERV29; SURF4; Surf-4; surf4 protein; surface 4 integral membrane protein; surfeit 4; surfeit gene 4; surfeit locus protein 4; zgc:65806; zgc:77007 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20932, 374273, 526045, 619346, 6836 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| A7YY49, O15260, Q64310, Q800K9 | |
| SURF4 | |
| Synthetic 18 amino acid peptide from N-Terminus of human SURF4. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Bovine, Chicken, Frog | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction