Learn More
sulfite oxidase, Mouse, Clone: 4F2, Abnova™
Description
Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins. [provided by RefSeq
Specifications
Specifications
| Antigen | sulfite oxidase |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 4F2 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant SUOX. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SUOX |
| Gene Accession No. | NM_000456 |
| Gene Symbols | SUOX |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.