Learn More
sulfatase modifying factor 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SUMF2.
Brand: Abnova H00025870-A01.50uL
Description
The catalytic sites of sulfatases are only active if they contain a unique amino acid, C-alpha-formylglycine (FGly). The FGly residue is posttranslationally generated from a cysteine by enzymes with FGly-generating activity. The gene described in this record is a member of the sulfatase-modifying factor family and encodes a protein with a DUF323 domain that localizes to the lumen of the endoplasmic reticulum. This protein has low levels of FGly-generating activity but can heterodimerize with another family member - a protein with high levels of FGly-generating activity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: QATSMVQLQGGRFLMGTNSPDSRDGEGPVREATVKPFAIDIFPVTNKDFRDFVREKKYRTEAEMFGWSFVFEDFVSDELRNKATQPMKSVLWWLPVEKAFSpecifications
| sulfatase modifying factor 2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SUMF2. | |
| SUMF2 | |
| DKFZp566I1024/DKFZp686I1024/DKFZp686L17160/DKFZp781L1035/MGC99485/pFGE | |
| Mouse | |
| 50 μL | |
| Yes | |
| 25870 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_015411 | |
| SUMF2 | |
| SUMF2 (NP_056226, 26 a.a. ∼ 125 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.