Learn More
SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human SUGT1 protein.
Brand: Abnova H00010910-D01P.100ug
Description
This gene is homologous to the yeast gene SGT1, which encodes a protein involved in kinetochore function and required for the G1/S and G2/M transitions. Complementation studies suggest that the human protein has similar functions. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MAAAAAGTATSQRFFQSFSDALIDEDPQAALEELTKALEQKPDDAQYYCQRAYCHILLGNYCVAVADAKKSLELNPNNSTAMLRKGICEYHEKNYAAALETFTEGQKLDSADANFSVWIKRCQEAQNGSESEVWTHQSKIKYDWYQTESQVVITLMIKNVQKNDVNVEFSEKELSALVKLPSGEDYNLKLELLHPIIPEQSTFKVLSTKIEIKLKKPEAVRWEKLEGQGDVPTPKQFVADVKNLYPSSSPYTRNWDKLVGEIKEEEKNEKLEGDAALNRLFQQIYSDGSDEVKRAMNKSFMESGGTVLSTNWSDVGKRKVEINPPDDMEWKKYSpecifications
| SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human SUGT1 protein. | |
| SUGT1 | |
| SGT1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_006704 | |
| SUGT1 | |
| SUGT1 (NP_006695.1, 1 a.a. ∼ 333 a.a) full-length human protein. | |
| 100 μg | |
| Cell Cycle | |
| 10910 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.