Learn More
syntaxin 11, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human STX11 protein.
Brand: Abnova H00008676-B01P.50ug
Description
This gene encodes a member of the syntaxin family. Syntaxins have been implicated in the targeting and fusion of intracellular transport vesicles. This family member may regulate protein transport among late endosomes and the trans-Golgi network. Mutations in this gene have been associated with familial hemophagocytic lymphohistiocytosis. [provided by RefSeq
Sequence: MKDRLAELLDLSKQYDQQFPDGDDEFDSPHEDIVFETDHILESLYRDIRDIQDENQLLVADVKRLGKQNARFLTSMRRLSSIKRDTNSIAKAIKARGEVIHCKLRAMKELSEAAEAQHGPHSAVARISRAQYNALTLTFQRAMHDYNQAEMKQRDNCKIRIQRQLEIMGKEVSGDQIEDMFEQGKWDVFSENLLADVKGARAALNEIESRHRELLRLESRIRDVHELFLQMAVLVEKQADTLNVIELNVQKTVDYTGQAKAQVRKAVQYEEKNPCRTLCCFCCPCLKSpecifications
| syntaxin 11 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human STX11 protein. | |
| STX11 | |
| FHL4/HLH4/HPLH4 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 8676 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_003764.2 | |
| STX11 | |
| STX11 (NP_003755.2, 1 a.a. ∼ 287 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.