missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptococcus agalactiae Monoclonal Antibody (BDI560), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA191193
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-91193 detects Streptococcus agalactiae from bacteria samples.
Once thought to infect only cows, where it produces mastitis, Streptococcus agalactiae is now known to be able to cause serious disease, bacteremia and meningitis, in immunocompromised individuals and in neonates. Infections in the newborn are the most common and are extremely serious. They are of two types. Early onset of infection, usually occuring within 5 days of birth, is manifested by bacteremia and pneumonia. This infection is contracted vertically, as the baby passes through the birth canal. The other type of infection is a meningitis that occurs between the 10th and 60th days of life. The origin of this infection is not known, but it is not believed to be the mother.Specifications
Streptococcus agalactiae | |
Monoclonal | |
0.78 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry | |
BDI560 | |
Unconjugated | |
s agalactiae; S. agalactiae | |
Tissue / cell preparation. UV-inactivated Strep. agalactiae cells isolated from mastitic milk. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |