missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STMN2, Mouse, Clone: 2E3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant STMN2.
Brand: Abnova H00011075-M15.100ug
This item is not returnable.
View return policy
Description
Superior cervical ganglion-10 is a neuronal growth-associated protein that shares significant amino acid sequence similarity with the phosphoprotein stathmin (MIM 151442).[supplied by OMIM
Sequence: SPKKKDLSLEEIQKKLEAAEERRKSQEAQVLKQLAEKREHEREVLQKALEENNNFSKMAEEKLILKMEQIKENREANLAAIIERLQEKERHAAEVRRNKELQVELSG*Specifications
| STMN2, | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_007029 | |
| STMN2 | |
| STMN2 (NP_008960, 73 a.a. ∼ 179 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2E3 | |
| Mouse monoclonal antibody raised against a partial recombinant STMN2. | |
| STMN2 | |
| SCG10/SCGN10/SGC10 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11075 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction