missing translation for 'onlineSavingsMsg'
Learn More
Learn More
serine/threonine kinase 40, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant STK40.
Brand: Abnova H00083931-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: PDIDDQMSNADSSQEAKVTEECSQYEFENYMRQQLLLAEEKSSIHDARSWVPKRQFGSAPPVRRLGHDAQPMTSLDTAILAQRYLRSpecifications
| serine/threonine kinase 40 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant STK40. | |
| STK40 | |
| MGC4796/RP11-268J15.4/SHIK/SgK495 | |
| Mouse | |
| 50 μL | |
| Kinases | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_032017 | |
| STK40 | |
| STK40 (NP_114406, 349 a.a. ∼ 434 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 83931 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction