missing translation for 'onlineSavingsMsg'
Learn More
Learn More
serine/threonine kinase 36, fused homolog (Drosophila), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human STK36 protein.
Brand: Abnova H00027148-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MEKYHVLEMIGEGSFGRVYKGRRKYSAQVVALKFIPKLGRSEKELRNLQREIEIMRGLRHPNIVHMLDSFETDKEVVVVTDYAEGELFQILEDDGKLPEDQVQAIAAQLVSALYYLHSHRILHRDMKPQNILLAKGGGIKLCDFGFARAMSTNTMVLTSIKGTPLYMSPELVEERPYDHTADLWSVGCILYELAVGTPPFYATSIFQLVSLILKDPVRWPSTISPCFKDFLQGLLTKDPRQRLSWPDLLYHPFIAGHVTIITEPAGPDLGTPFTSRLPPELQVLKDEQAHRLAPKGNQSRILTQAYKRMAEEAMQKKHQNTGPALEQEDKTSKVAPGTAPLPRLGATPQESSLLAGILASELKSSWAKSGTGEVPSAPRENRTTPDCERAFPEERPEVLGQRSTDVVDLENEEPDSDNEWQHLLETTEPVPIQLKAPLTLLCNPDFCQRIQSQLHEAGGQILNGILEGASHILPAFRVLSSLLSSCSDSVALYSFCREAGLPGLLLSLLRHSQESNSLQQQSWYGTFLQDLMAVIQAYFACTFNLERSQTSDSLQVFQEAANLFLDLLGKLLAQPDDSEQTLRRDSLMCFTVLCEAMDGNSRAISKAFYSSLLTTQQVVLDGLLHGLTVPQLPVHTPQGAPQVSQPLREQSEDIPGAISSALAAICTAPVGLPDCWDAKEQVCWHLANQLTEDSSQLRPSLISGLQHPILCLHLLKVLYSCCLVSEGLCRLLGQEPLALESLFMLIQGKVKVVDWEESTEVTLYFLTLLVFRLQNLPCGMEKLGSDVATLFTHSHVVSLVSAAACLLGQLGQQGVTFDLQPMEWMAAATHALSAPAEVRLTPPGSCGFYDGLLILLLQLLTEQGKASLIRDMSSSEMWTVLWHRFSMVLRLPEEASAQEGELSLSSPPSPEPDWTLISPQGMAALLSLAMATFTQEPQLCLSCLSQHGSILMSILKHLLCPSFLNQLRQAPHGSEFLPVVVLSVCQLLCFPFALDMDADLLIGVLADLRDSEVAAHLLQVCCYHLPLMQVELPISLLTRLALMDPTSLNQFVNTVSASPRTIVSFLSVALLSDQPLLTSDLLSLLAHTARVLSPSHLSFIQELLAGSDESYRPLRSLLGHPENSVRAHTYRLLGHLLQHSMALRGALQSQSGLLSLLLLGLGDKDPVVRCSASFAVGNAAYQAGPLGPALAAAVPSMTQLLGDPQAGIRRNVASALGNLGPEGLGEELLQCEVPQRLLEMACGDPQPNVKEAALIALRSLQQEPGIHQVLVSLGASEKLSLLSLGNQSLPHSSPRPASAKHCRKLIHLLRPAHSMSpecifications
| serine/threonine kinase 36, fused homolog (Drosophila) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human STK36 protein. | |
| STK36 | |
| DKFZp434N0223/FU/KIAA1278 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_015690.2 | |
| STK36 | |
| STK36 (NP_056505.1, 1 a.a. ∼ 1315 a.a) full-length human protein. | |
| 50 μg | |
| Kinases | |
| 27148 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction