missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531551
This item is not returnable.
View return policy
Description
Recommended positive controls: Stk31 tansfected AZ521 cell. Predicted reactivity: Mouse (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is similar to a mouse gene that encodes a putative protein kinase with a tudor domain, and shows testis-specific expression. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| STK31 | |
| Polyclonal | |
| Unconjugated | |
| STK31 | |
| C330007K24Rik; RGD1564384; serine threonine kinase 31; serine/threonine kinase 31; serine/threonine-protein kinase 31; Serine/threonine-protein kinase NYD-SPK; SGK396; STK31; Sugen kinase 396; TDRD8; testis tissue sperm-binding protein Li 78m; tudor domain containing 8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56164, 77485 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q99MW1, Q9BXU1 | |
| STK31 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of mouse Stk31. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction