missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529415
This item is not returnable.
View return policy
Description
Recommended positive controls: U87-MG. Predicted reactivity: Mouse (93%), Rat (95%), Rhesus Monkey (98%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
STK16 (PKL12) is a serine/threonine kinase that functions in signal transduction pathways and transcriptional regulation. Protein kinase that act on both serine and threonine residues. STK16 may regulate transforming growth factor-beta (Tgfb) and vascular endothelial growth factor transcription.
Specifications
| STK16 | |
| Polyclonal | |
| Unconjugated | |
| STK16 | |
| Edpk; Embryo-derived protein kinase; F52; hPSK; kinase related to cerevisiae and thaliana; Krct; MPSK; Mpsk1; Myristoylated and palmitoylated serine/threonine-protein kinase; PKL12; protein kinase expressed in day 12 fetal liver; Protein kinase Krct; Protein kinase PKL12; serine/threonine kinase 16; serine/threonine-protein kinase 16; Stk16; TGF-beta-stimulated factor 1; Tsf1; TSF-1; tyrosine-protein kinase STK16 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8576 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O75716 | |
| STK16 | |
| Recombinant protein encompassing a sequence within the center region of human STK16. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction