missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAU2 Monoclonal Antibody (3B7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521686
This item is not returnable.
View return policy
Description
Peptide Sequence: LGYKASTNLQ DQLEKTGENK GWSGPKPGFP EPTNNTPKGI LHLSPDVYQE MEASRHKVIS GTTLGYLSPK DMNQPSSSFF SISPTSNSSA TIARELLMNG.
Staufen homolog 2 is a member of the family of double-stranded RNA (dsRNA)-binding proteins involved in the transport and/or localization of mRNAs to different subcellular compartments and/or organelles. These proteins are characterized by the presence of multiple dsRNA-binding domains which are required to bind RNAs having double-stranded secondary structures. Staufen homolog 2 shares 48. 5% and 59. 9% similarity with drosophila and human staufen, respectively. The exact function of Staufen homolog 2 is not known, but since it contains 3 copies of conserved dsRNA binding domain, it could be involved in double-stranded RNA binding events. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| STAU2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9NUL3 | |
| Stau2 | |
| STAU2 (NP_055208, 341 a.a. ~ 440 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 3B7 | |
| Unconjugated | |
| Stau2 | |
| 39K2; 39K3; Double-stranded RNA-binding protein Staufen homolog 2; r-staufen protein; Stau2; staufen (RNA binding protein) homolog 2 (Drosophila); staufen 2; staufen double-stranded RNA binding protein 2; staufen homolog 2; staufen, RNA binding protein, homolog 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 27067 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction