Learn More
PSF Progressive Safety™ Anti-static Safety Wellington Boots To EN345
Product Code. 12772096
Click to view available options
Size:
3
4
5
6
7
8
9
10
11
12
13
Unit Size:
1 pair
Product Code. 12772096
Brand: PSF Progressive Safety™ AGRIP/W4
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
  • Antislip
  • Anti-static
  • Waterproof rubber sole
  • Resistant to heat, oils, acids & animal fats
  • 200 joule toe protection
  • White
Specifications
Size 4